• USA Home
  • Search Results

  • 1phenyldodecane2464312301311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Phenyldodecane

3 Products
Synonym:   Dodecyl benzene, Dodecyl benzene
Image of 1-Phenyldodecane
CAS Number:
Linear Formula:
 C6H5(CH2)11CH3
Molecular Weight:
 246.43
 EC Index Number
 204-591-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More