• USA Home
  • Search Results

  • 1phenylpiperazine162239254611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Phenylpiperazine

1 Products
Synonym:   N-Phenyldiethylenediamine
Image of 1-Phenylpiperazine
CAS Number:
Empirical Formula (Hill Notation):
 C10H14N2
Molecular Weight:
 162.23
 Beilstein/REAXYS Number:
 132157
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P30004 1-Phenylpiperazine, 99%,