• USA Home
  • Search Results

  • 1propanol60107123811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Propanol

12 Products
Synonym:   1-Propanol, n-Propyl alcohol, Propan-1-ol
Image of 1-Propanol
CAS Number:
Empirical Formula (Hill Notation):
 C3H8O
Molecular Weight:
 60.10
 EC Index Number
 200-746-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Restriction
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Color
  Anal. Standard Form
  Special Grade
  pH Value
  UV-Absorption Condition
  Transition temperature
  Purity Grade
  Density
  Autoignition temp
  Vapor pressure
  Melting Point
  Explosion limit
  Solubility
  Transmittance
  Physical Form
  UV-Absorption Values
  Assay
Show More