• USA Home
  • Search Results

  • 1pyrenebutyricacid28834344345611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Pyrenebutyric acid

1 Products
Synonym:   4-(1-Pyrenyl)butyric acid, PyBA
Image of 1-Pyrenebutyric acid
CAS Number:
Empirical Formula (Hill Notation):
 C20H16O2
Molecular Weight:
 288.34
 Beilstein/REAXYS Number:
 2140554
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Fluorescence Test
Show More

Product #
Description
257354 1-Pyrenebutyric acid, 97%,