• USA Home
  • Search Results

  • 1pyrenecarboxylicacid246261969402111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Pyrenecarboxylic acid

1 Products
Image of 1-Pyrenecarboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C17H10O2
Molecular Weight:
 246.26
 Beilstein/REAXYS Number:
 2375854
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More