• USA Home
  • Search Results

  • 1r2r12dicyclohexyl12ethanediol2263612085092211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1R,2R)-1,2-Dicyclohexyl-1,2-ethanediol

1 Products
Synonym:   (1R,2R)-1,2-Dicyclohexylethylene glycol
Image of (1<I>R</I>,2<I>R</I>)<WBR>-<WBR>1,2-<WBR>Dicyclohexyl-<WBR>1,2-<WBR>ethanediol
CAS Number:
Linear Formula:
 [C6H11CH(OH)-]2
Molecular Weight:
 226.36
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More