• USA Home
  • Search Results

  • 1r2s2amino12diphenylethanol213282319016111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1R,2S)-(−)-2-Amino-1,2-diphenylethanol

1 Products
Image of (1<I>R</I>,2<I>S</I>)-(−)-2-Amino-1,2-diphenylethanol
CAS Number:
Linear Formula:
 C6H5CH(NH2)CH(C6H5)OH
Molecular Weight:
 213.28
 Beilstein/REAXYS Number:
 2806218
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More