• USA Home
  • Search Results

  • 1r2sephedrinehydrochloride201695098611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1R,2S)-(−)-Ephedrine hydrochloride

1 Products
Synonym:   (1R,2S)-α-[1-(Methylamino)ethyl]benzyl alcohol hydrochloride, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol hydrochloride, (1R,2S)-(−)-Ephedrine hydrochloride
Image of (1R,2S)-(−)-Ephedrine hydrochloride
CAS Number:
Linear Formula:
 C6H5CH[CH(NHCH3)CH3]OH · HCl
Molecular Weight:
 201.69
 Beilstein/REAXYS Number:
 3630391
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More