• USA Home
  • Search Results

  • 1s2rDephedrined3hydrochloridesolution2047128597973911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1S,2R)-(+)-Ephedrine-d3 hydrochloride solution

2 Products
Image of (1S,2R)<WBR>-<WBR>(+)<WBR>-<WBR>Ephedrine-<WBR>d<SUB>3</SUB> hydrochloride solution
CAS Number:
Empirical Formula (Hill Notation):
 C10H13ClNOD3
Molecular Weight:
 204.71
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More