• USA Home
  • Search Results

  • 1s2rDephedrinehydrochloride201692422186111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1S,2R)-(+)-Ephedrine hydrochloride

2 Products
Synonym:   (1S,2R)-(+)-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride, (1S,2R)-(+)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride
Image of (1<I>S</I>,2<I>R</I>)-(+)-Ephedrine hydrochloride
CAS Number:
Linear Formula:
 C6H5CH[CH(NHCH3)CH3]OH · HCl
Molecular Weight:
 201.69
 Beilstein/REAXYS Number:
 6118541
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More