• USA Home
  • Search Results

  • 1s2s4rDlimonene12diol170253863075011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1S,2S,4R)-(+)-Limonene-1,2-diol

1 Products
Synonym:   (+)-(1S,2S,4R)-Limonene glycol, (+)-1-Hydroxyneodihydrocarveol, (1S,2S,4R)-8-p-Menth-8-ene-1,2-diol, (1S,2S,4R)-(+)-4-Isopropenyl-1-methylcyclohexan-1,2-diol, (1S,2S,4R)-(+)-p-Menth-8-en-1,2-diol
Image of (1<I>S</I>,2<I>S</I>,4<I>R</I>)<WBR>-<WBR>(+)<WBR>-<WBR>Limonene-<WBR>1,2-<WBR>diol
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O2
Molecular Weight:
 170.25
 Beilstein/REAXYS Number:
 2325340
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More