• USA Home
  • Search Results

  • 1s2sD2amino14nitrophenyl13propanediol21220296448911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1S,2S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

1 Products
Synonym:   L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Image of (1<I>S</I>,2<I>S</I>)<WBR>-<WBR>(+)<WBR>-<WBR>2-<WBR>Amino-<WBR>1-<WBR>(4-<WBR>nitrophenyl)<WBR>-<WBR>1,3-<WBR>propanediol
CAS Number:
Linear Formula:
 O2NC6H4CH(OH)CH(NH2)CH2OH
Molecular Weight:
 212.20
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More