• USA Home
  • Search Results

  • 1s2sDpseudoephedrine165239082411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(1S,2S)-(+)-Pseudoephedrine

1 Products
Synonym:   (+)-ψ-Ephedrine, (+)-Pseudoephedrine, (1S,2S)-2-Methylamino-1-phenyl-1-propanol, d-Isoephedrine, d-Pseudoephedrine
Image of (1<I>S</I>,2<I>S</I>)-(+)-Pseudoephedrine
CAS Number:
Empirical Formula (Hill Notation):
 C10H15NO
Molecular Weight:
 165.23
 Beilstein/REAXYS Number:
 2414132
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More