• USA Home
  • Search Results

  • 1s9rbicucullinemethiodide509294070969111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1(S),9(R)-(−)-Bicuculline methiodide

1 Products
Synonym:   (5S)-5-[(6R)-6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl]-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium iodide
Image of 1(<I>S</I>),9(<I>R</I>)-(−)-Bicuculline methiodide
CAS Number:
Empirical Formula (Hill Notation):
 C21H20INO6
Molecular Weight:
 509.29
 Beilstein/REAXYS Number:
 5419649
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More