• USA Home
  • Search Results

  • 1stearoylracglycerol3585612394411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Stearoyl-rac-glycerol

2 Products
Synonym:   1-Monooctadecanoyl-rac-glycerol, rac-Glycerol 1-stearate, DL-α-Stearin, MG(18:0/0:0/0:0)[rac], Monostearin
Image of 1-Stearoyl-<I>rac</I>-glycerol
CAS Number:
Linear Formula:
 CH3(CH2)16COOCH2CH(OH)CH2OH
Molecular Weight:
 358.56
 Beilstein/REAXYS Number:
 1728685
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More