• USA Home
  • Search Results

  • 1thioglycerol108169627511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Thioglycerol

5 Products
Synonym:   α-Monothioglycerol, α-Thioglycerol, 3-Mercapto-1,2-propanediol
Image of 1-Thioglycerol
CAS Number:
Linear Formula:
 HSCH2CH(OH)CH2OH
Molecular Weight:
 108.16
 Beilstein/REAXYS Number:
 1732046
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Physical Form
  Suitability
  Product Line
Show More