• USA Home
  • Search Results

  • 1triacontanol4388159350011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Triacontanol

1 Products
Synonym:   1-Hydroxytriacontane, Melissyl alcohol
Image of 1-Triacontanol
CAS Number:
Linear Formula:
 CH3(CH2)28CH2OH
Molecular Weight:
 438.81
 Beilstein/REAXYS Number:
 1711965
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More