• USA Home
  • Search Results

  • 1trimethylsiloxy13butadiene14227665143011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(Trimethylsiloxy)-1,3-butadiene

1 Products
Image of 1-<WBR>(Trimethylsiloxy)<WBR>-<WBR>1,3-<WBR>butadiene
CAS Number:
Linear Formula:
 (CH3)3SiOCH=CHCH=CH2
Molecular Weight:
 142.27
 Beilstein Registry Number:
 1902024
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More