• USA Home
  • Search Results

  • 1trimethylsilyl1hexyne15432384494811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Trimethylsilyl-1-hexyne

1 Products
Image of 1-<WBR>Trimethylsilyl-<WBR>1-<WBR>hexyne
CAS Number:
Linear Formula:
 CH3(CH2)3C≡CSi(CH3)3
Molecular Weight:
 154.32
 Beilstein/REAXYS Number:
 1421652
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More