• USA Home
  • Search Results

  • 1trimethylsilylpropyne11224622491511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-(Trimethylsilyl)propyne

1 Products
Synonym:   Trimethyl-1-propynylsilane
Image of 1-<WBR>(Trimethylsilyl)<WBR>propyne
CAS Number:
Linear Formula:
 CH3C≡CSi(CH3)3
Molecular Weight:
 112.24
 Beilstein/REAXYS Number:
 1071311
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More