• USA Home
  • Search Results

  • 1vinyl2pyrrolidinone111148812011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1-Vinyl-2-pyrrolidinone

2 Products
Synonym:   1-Vinyl-2-pyrrolidone
Image of 1-<WBR>Vinyl-<WBR>2-<WBR>pyrrolidinone
CAS Number:
Empirical Formula (Hill Notation):
 C6H9NO
Molecular Weight:
 111.14
 Beilstein/REAXYS Number:
 110513
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
  Assay
  Special Grade
Show More