• USA Home
  • Search Results

  • 2223tetrafluoroacetophenone1921170864511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2,2,3′-Tetrafluoroacetophenone

1 Products
Synonym:   α,α,α,3-Tetrafluoroacetophenone
Image of 2,2,2,3′-Tetrafluoroacetophenone
CAS Number:
Linear Formula:
 FC6H4COCF3
Molecular Weight:
 192.11
 Beilstein/REAXYS Number:
 1872788
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More