• USA Home
  • Search Results

  • 2224tetrafluoroacetophenone1921165532311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2,2,4′-Tetrafluoroacetophenone

1 Products
Synonym:   α,α,α,4-Tetrafluoroacetophenone
Image of 2,2,2,4′-Tetrafluoroacetophenone
CAS Number:
Linear Formula:
 FC6H4COCF3
Molecular Weight:
 192.11
 Beilstein/REAXYS Number:
 1872824
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More