• USA Home
  • Search Results

  • 22333pentafluoro1propanol1500542205911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2,3,3,3-Pentafluoro-1-propanol

2 Products
Synonym:   PFPOH
Image of 2,2,3,3,3-<WBR>Pentafluoro-<WBR>1-<WBR>propanol
CAS Number:
Linear Formula:
 CF3CF2CH2OH
Molecular Weight:
 150.05
 Beilstein/REAXYS Number:
 1743133
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More