• USA Home
  • Search Results

  • 224trimethyl13pentanedioldiisobutyrate28641684650011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate

2 Products
Synonym:   1-Isopropyl-2,2-dimethyltrimethylene diisobutyrate
Image of 2,2,4-<WBR>Trimethyl-<WBR>1,3-<WBR>pentanediol diisobutyrate
CAS Number:
Linear Formula:
 (CH3)2CHCH[O2CCH(CH3)2]C(CH3)2CH2O2CCH(CH3)2
Molecular Weight:
 286.41
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More