• USA Home
  • Search Results

  • 2262terpyridine23327114879411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2′:6′,2′′-Terpyridine

2 Products
Synonym:   α,α′,α″-Tripyridyl, 2,6-Di(2-pyridyl)pyridine
Image of 2,2′:6′,2′′-Terpyridine
CAS Number:
Empirical Formula (Hill Notation):
 C15H11N3
Molecular Weight:
 233.27
 Beilstein/REAXYS Number:
 11199
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More