• USA Home
  • Search Results

  • 22azobis2methylpropionitrile164217867111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2′-Azobis(2-methylpropionitrile)

2 Products
Synonym:   α,α′-Azoisobutyronitrile, AIBN, Azobisisobutyronitrile, Free radical initiator
Image of 2,2′-Azobis(2-methylpropionitrile)
CAS Number:
Linear Formula:
 (CH3)2C(CN)N=NC(CH3)2CN
Molecular Weight:
 164.21
 Beilstein/REAXYS Number:
 1708400
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More