• USA Home
  • Search Results

  • 22azobis2methylpropionitrilesolution164217867111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2′-Azobis(2-methylpropionitrile) solution

1 Products
Synonym:   α,α,′-Azoisobutyronitrile solution, AIBN solution
Image of 2,2′-Azobis(2-methylpropionitrile) solution
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N4
Molecular Weight:
 164.21
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More