• USA Home
  • Search Results

  • 22dimethoxypropane104157776911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2-Dimethoxypropane

3 Products
Synonym:   Acetone dimethyl acetal
Image of 2,2-Dimethoxypropane
CAS Number:
Linear Formula:
 (CH3)2C(OCH3)2
Molecular Weight:
 104.15
 Beilstein/REAXYS Number:
 635678
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
  Application(s)
  Potency
Show More