• USA Home
  • Search Results

  • 22dimethyl36912tetraoxa2silatridecane2363886407962911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,2-Dimethyl-3,6,9,12-tetraoxa-2-silatridecane

1 Products
Synonym:   ANL-1NM3
Image of 2,2-<WBR>Dimethyl-<WBR>3,6,9,12-<WBR>tetraoxa-<WBR>2-<WBR>silatridecane
CAS Number:
Empirical Formula (Hill Notation):
 C10H24O4Si
Molecular Weight:
 236.38
 MDL number:
 NACRES:
 NA.23
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More