• USA Home
  • Search Results

  • 23456pentabromoethylbenzene500658522311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,4,5,6-Pentabromoethylbenzene

1 Products
Synonym:   Pentabromoethylbenzene
Image of 2,3,4,5,6-<WBR>Pentabromoethylbenzene
CAS Number:
Empirical Formula (Hill Notation):
 C8H5Br5
Molecular Weight:
 500.65
 Beilstein Registry Number:
 3133073
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More