• USA Home
  • Search Results

  • 23456pentafluorobenzaldehyde1960765337211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,4,5,6-Pentafluorobenzaldehyde

1 Products
Image of 2,3,4,5,6-<WBR>Pentafluorobenzaldehyde
CAS Number:
Linear Formula:
 C6F5CHO
Molecular Weight:
 196.07
 Beilstein/REAXYS Number:
 1876550
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More