• USA Home
  • Search Results

  • 2346tetrachlorophenolsolution231895890211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,4,6-Tetrachlorophenol solution

1 Products
Image of 2,3,4,6-<WBR>Tetrachlorophenol solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H2Cl4O
Molecular Weight:
 231.89
 Beilstein/REAXYS Number:
 779754
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Concentration
Show More