• USA Home
  • Search Results

  • 2356tetrafluoro4trifluoromethylbenzylbromide310997643740611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide

1 Products
Synonym:   1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide
Image of 2,3,5,6-<WBR>Tetrafluoro-<WBR>4-<WBR>(trifluoromethyl)<WBR>benzyl bromide
CAS Number:
Linear Formula:
 CF3C6F4CH2Br
Molecular Weight:
 310.99
 Beilstein/REAXYS Number:
 8423469
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More