• USA Home
  • Search Results

  • 235tribromothiophene32083314124011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,5-Tribromothiophene

1 Products
Image of 2,3,5-<WBR>Tribromothiophene
CAS Number:
Empirical Formula (Hill Notation):
 C4HBr3S
Molecular Weight:
 320.83
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More