• USA Home
  • Search Results

  • 235trimethylpyrazine122171466755111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3,5-Trimethylpyrazine

4 Products
Synonym:   Potato pyrazine
Image of 2,3,5-<WBR>Trimethylpyrazine
CAS Number:
Empirical Formula (Hill Notation):
 C7H10N2
Molecular Weight:
 122.17
 Council of Europe no.:
 735
 EC Number:
 FEMA Number:
 3244
 Flavis number:
 14.019
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More