• USA Home
  • Search Results

  • 23benzofuran1181327189611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Benzofuran

2 Products
Synonym:   Coumarone
Image of 2,3-Benzofuran
CAS Number:
Empirical Formula (Hill Notation):
 C8H6O
Molecular Weight:
 118.13
 Beilstein/REAXYS Number:
 107704
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More