• USA Home
  • Search Results

  • 23butanediol901251385911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Butanediol

3 Products
Synonym:   2,3-Butylene glycol
Image of 2,3-Butanediol
CAS Number:
Linear Formula:
 CH3CH(OH)CH(OH)CH3
Molecular Weight:
 90.12
 Beilstein/REAXYS Number:
 969165
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More