• USA Home
  • Search Results

  • 23butanedione860943103811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Butanedione

2 Products
Synonym:   Biacetyl, Diacetyl
Image of 2,3-Butanedione
CAS Number:
Linear Formula:
 CH3COCOCH3
Molecular Weight:
 86.09
 Beilstein/REAXYS Number:
 605398
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More