• USA Home
  • Search Results

  • 23diaminonaphthalene1582077197111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Diaminonaphthalene

2 Products
Synonym:   2,3-Naphthalenediamine, DAN
Image of 2,3-<WBR>Diaminonaphthalene
CAS Number:
Empirical Formula (Hill Notation):
 C10H10N2
Molecular Weight:
 158.20
 Beilstein/REAXYS Number:
 2206394
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More