• USA Home
  • Search Results

  • 23dichloroquinoxaline19904221363011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Dichloroquinoxaline

1 Products
Image of 2,3-<WBR>Dichloroquinoxaline
CAS Number:
Empirical Formula (Hill Notation):
 C8H4Cl2N2
Molecular Weight:
 199.04
 Beilstein/REAXYS Number:
 126076
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More