• USA Home
  • Search Results

  • 23dideoxyuridine21220598309511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′,3′-Dideoxyuridine

1 Products
Synonym:   ddU
Image of 2′,3′-Dideoxyuridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H12N2O4
Molecular Weight:
 212.20
 Beilstein/REAXYS Number:
 750592
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More