• USA Home
  • Search Results

  • 23dihydro5methylfuran8412148715611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Dihydro-5-methylfuran

1 Products
Image of 2,3-<WBR>Dihydro-<WBR>5-<WBR>methylfuran
CAS Number:
Empirical Formula (Hill Notation):
 C5H8O
Molecular Weight:
 84.12
 Beilstein/REAXYS Number:
 103487
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More