• USA Home
  • Search Results

  • 23dihydroxybenzaldehyde138122467778911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Dihydroxybenzaldehyde

1 Products
Image of 2,3-<WBR>Dihydroxybenzaldehyde
CAS Number:
Linear Formula:
 (HO)2C6H3CHO
Molecular Weight:
 138.12
 Beilstein Registry Number:
 2041781
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More