• USA Home
  • Search Results

  • 23dimethoxy5methylpbenzoquinone1821760594711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Dimethoxy-5-methyl-p-benzoquinone

1 Products
Synonym:   Coenzyme Q0
Image of 2,3-<WBR>Dimethoxy-<WBR>5-<WBR>methyl-<I>p</I>-benzoquinone
CAS Number:
Empirical Formula (Hill Notation):
 C9H10O4
Molecular Weight:
 182.17
 Beilstein/REAXYS Number:
 1640422
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More