• USA Home
  • Search Results

  • 23hexanedione11414384824611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Hexanedione

5 Products
Synonym:   propyl diketone, methylpropylglyoxal
Image of 2,3-Hexanedione
CAS Number:
Empirical Formula (Hill Notation):
 C6H10O2
Molecular Weight:
 114.14
 EC Index Number
 223-350-8
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Solubility
  Assay
  Special Grade
Show More