• USA Home
  • Search Results

  • 23naphthalenedicarboxaldehyde18419714949711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,3-Naphthalenedicarboxaldehyde

1 Products
Synonym:   NDA
Image of 2,3-<WBR>Naphthalenedicarboxaldehyde
CAS Number:
Linear Formula:
 C10H6(CHO)2
Molecular Weight:
 184.19
 Beilstein/REAXYS Number:
 2207267
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More