• USA Home
  • Search Results

  • 23oisopropylideneuridine2842736243611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2′,3′-O-Isopropylideneuridine

1 Products
Image of 2′,3′-<I>O</I>-Isopropylideneuridine
CAS Number:
Empirical Formula (Hill Notation):
 C12H16N2O6
Molecular Weight:
 284.27
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More