• USA Home
  • Search Results

  • 23tetrahydrofurfuryloxytetrahydropyran1862571014511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2(3)-(Tetrahydrofurfuryloxy)tetrahydropyran

1 Products
Synonym:   ‘T-Solvent’
Image of 2(3)<WBR>-<WBR>(Tetrahydrofurfuryloxy)<WBR>tetrahydropyran
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O3
Molecular Weight:
 186.25
 Beilstein/REAXYS Number:
 109779
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More