• USA Home
  • Search Results

  • 245trimethylanilinesolution1352113717711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

2,4,5-Trimethylaniline solution

1 Products
Image of 2,4,5-<WBR>Trimethylaniline solution
CAS Number:
Empirical Formula (Hill Notation):
 C9H13N
Molecular Weight:
 135.21
 Beilstein/REAXYS Number:
 2352436
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More